Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD2 Mouse anti-Human, Clone: PCRP-GTF2IRD2-1B12, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen GTF2IRD2
Clone PCRP-GTF2IRD2-1B12
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Applications Western Blot, Flow Cytometry, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122272
SDP
View Documents
Novus Biologicals
NBP30739920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

GTF2IRD2 Monoclonal specifically detects GTF2IRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
Specifications

Specifications

GTF2IRD2
0.2 mg/ml
Western Blot, Flow Cytometry, Peptide Array
Unconjugated
Mouse
Human
FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha
Recombinant full-length human GTF2IRD2 protein
Primary
Store at 4°C.
PCRP-GTF2IRD2-1B12
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
84163
IgG2b
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.