Learn More
GTF2IRD2 Mouse anti-Human, Clone: PCRP-GTF2IRD2-1B4, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | GTF2IRD2 |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
| Classification | Monoclonal |
| Clone | PCRP-GTF2IRD2-1B4 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.