Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2IRD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566031 Supplier Invitrogen™ Supplier No. PA566031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPTQKSPVQP VEDLAGNLWE KLREKIRSFV AYSIAIDEIT DINNTTQLAI FIRGVDENFD VSEELLDTVP MTGT Highest antigen sequence identity to the following orthologs - mouse 83%, rat 29%.

This gene encodes a glycosylated phosphoprotein with a leucine zipper motif, two helix-loop-helix motifs (I repeats) that are similar to domains found in the TFII-I family of transcription factors, one CHARLIE8 transposable element-like sequence, and a BED zinc finger. This gene lies within a region that is deleted in Williams-Beuren syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2IRD2B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF2IRD2B
Gene Accession No. Q6EKJ0
Gene Alias general transcription factor 2 I repeat domain-containing 2 beta; general transcription factor II-I repeat domain-containing protein 2B; GTF2I repeat domain containing 2B; GTF2I repeat domain-containing protein 2B; GTF2IRD2 beta; GTF2IRD2B; Transcription factor GTF2IRD2-beta
Gene Symbols GTF2IRD2B
Host Species Rabbit
Immunogen Recombinant Human GTF2IRD2B. Recombinant protein control fragment (Product #RP-105629).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389524
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.