Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD2B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549213

Catalog No. PA549213

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a glycosylated phosphoprotein with a leucine zipper motif, two helix-loop-helix motifs (I repeats) that are similar to domains found in the TFII-I family of transcription factors, one CHARLIE8 transposable element-like sequence, and a BED zinc finger. This gene lies within a region that is deleted in Williams-Beuren syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq, Jul 2008]
Specifications

Specifications

GTF2IRD2B
Polyclonal
PBS with 0.09% sodium azide
Q6EKJ0
General transcription factor II-I repeat domain-containing protein 2B, GTF2I repeat domain-containing protein 2B, Transcription factor GTF2IRD2-beta, GTF2IRD2B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389524
Western Blot
Unconjugated
GTF2IRD2B
Liquid
GTF2IRD2B
KLH conjugated synthetic peptide between 467-494 amino acids from the C-terminal region of human GTF2IRD2B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit