Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF3A Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204017AF350
Description
GTF3A Polyclonal antibody specifically detects GTF3A in Human samples. It is validated for Western BlotSpecifications
GTF3A | |
Polyclonal | |
Western Blot | |
general transcription factor IIIA, TFIIIAAP2, transcription factor IIIA | |
The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_002088.2 (GeneID 2971). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
2971 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction