Learn More
Invitrogen™ GTF3A Monoclonal Antibody (4B4)

Description
Peptide Sequence: NQQKQYICSF EDCKKTFKKH QQLKIHQCQN TNEPLFKCTQ EGCGKHFASP SKLKRHAKAH EGYVCQKGCS FVAKTWTELL KHVRETHKEE.
Specifications
Specifications
| Antigen | GTF3A |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4B4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GTF3A |
| Gene Accession No. | Q92664 |
| Gene Alias | AP2; general transcription factor IIIA; GTF3A; TF3A; TFIIIA; Transcription factor IIIA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.