Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF3A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA552497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552497 Supplier Thermo Scientific Supplier No. PA552497

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: KRKDYLKQHM KTHAPERDVC RCPREGCGRT YTTVFNLQSH ILSFHEESRP FVCEHAGCGK TFAMKQSLTR HAVVHDPDKK KMKLKVKKSR EKRSLASHLS GYIPPKRKQG QGLSLCQNGE SPNCVEDKML STV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.

The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50 bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3A
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF3A
Gene Accession No. Q92664
Gene Alias AP2; general transcription factor IIIA; GTF3A; TF3A; TFIIIA; Transcription factor IIIA
Gene Symbols GTF3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF3A
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.