Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550146

Catalog No. PA550146


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total TFIIIA / GTF3A protein.

The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50 bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GTF3A
Polyclonal
Unconjugated
GTF3A
AP2; general transcription factor IIIA; GTF3A; TF3A; TFIIIA; Transcription factor IIIA
Rabbit
Antigen Affinity Chromatography
RUO
2971
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q92664
GTF3A
Synthesized peptide derived from Internal of human GTF3A.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.