Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562441 Supplier Invitrogen™ Supplier No. PA562441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVFIFLKKNA VIVDTTICDP HYNLARSSRP FERRLYVLNS MQDVENYWFD LQCVCLNTPL GVVRCPRVRK NSSTDQGSDE EGSLQKEQES AMDKHNLERK CAMLEYTTGS REVVDEGLIP GDGLGAAGLD SSFYG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

GTF3C1 (General Transcription Factor IIIC Subunit 1) is a Protein Coding gene. Diseases associated with GTF3C1 includeNonparalytic PoliomyelitisandBulbar Polio. Among its related pathways areActivated PKN1 stimulates transcription of AR (androgen receptor) regulated genes KLK2 and KLK3andGene Expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3C1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF3C1
Gene Accession No. Q12789
Gene Alias DKFZp686A111; general transcription factor 3C polypeptide 1; general transcription factor IIIC subunit 1; general transcription factor IIIC, polypeptide 1 (alpha subunit); general transcription factor IIIC, polypeptide 1, alpha 220kDa; GTF3C1; TF3C-alpha; TFIIIC; TFIIIC 220 kDa subunit; TFIIIC box B-binding subunit; TFIIIC220; TFIIICalpha; transcription factor IIIC 220 kDa subunit; Transcription factor IIIC subunit alpha
Gene Symbols GTF3C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF3C1. Recombinant protein control fragment (Product #RP-102471).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.