Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF3C1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140181
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PVDIVFERDM LTQTYDLIER RGTKGISQAE IRVAMNVGKL EARMLCRLLQ RFKVVKGFME DEGRQRTTKY ISCVFAEESD LSRQYQREKA RSELLTTVS.
GTF3C1 (General Transcription Factor IIIC Subunit 1) is a Protein Coding gene. Diseases associated with GTF3C1 includeNonparalytic PoliomyelitisandBulbar Polio. Among its related pathways areActivated PKN1 stimulates transcription of AR (androgen receptor) regulated genes KLK2 and KLK3andGene Expression.
Specifications
GTF3C1 | |
Polyclonal | |
Unconjugated | |
GTF3C1 | |
DKFZp686A111; general transcription factor 3C polypeptide 1; general transcription factor IIIC subunit 1; general transcription factor IIIC, polypeptide 1 (alpha subunit); general transcription factor IIIC, polypeptide 1, alpha 220kDa; GTF3C1; TF3C-alpha; TFIIIC; TFIIIC 220 kDa subunit; TFIIIC box B-binding subunit; TFIIIC220; TFIIICalpha; transcription factor IIIC 220 kDa subunit; Transcription factor IIIC subunit alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12789 | |
GTF3C1 | |
Recombinant protein corresponding to Human GTF3C1. Recombinant protein control fragment (Product #RP-109511). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction