Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF3C1/TFIIIC220 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen GTF3C1/TFIIIC220
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-0004
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301293AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).
Specifications

Specifications

GTF3C1/TFIIIC220
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
DKFZp686A111, TFIIIC, TFIIIC220, TFIIICalpha, TF3C-alpha; TFIIIC 220 kDa subunit; TFIIIC box B-binding subunit; general transcription factor 3C polypeptide 1; general transcription factor IIIC, polypeptide 1 (alpha subunit); transcription factor IIIC 220 kDa subunit; transcription factor IIIC subunit alpha
GTF3C1
IgG
Antigen affinity chromatography
Antibody
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human
Q12789
2975
Between 2059 and 2109
Primary
4° C
GTF3C1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.