Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF3C1/TFIIIC220 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560002
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0002 Supplier Bethyl Laboratories, Inc Supplier No. A301293A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

SIII, also known as elongin, is a general transcription elongation factor that increases the RNA polymerase II transcription elongation past template-encoded arresting sites. Subunit A is transcriptionally active and its transcription activity is strongly enhanced by binding to the dimeric complex of the SIII regulatory subunits B and C (elongin BC complex).

Specifications

Antigen GTF3C1/TFIIIC220
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene GTF3C1
Gene Accession No. Q12789
Gene Alias DKFZp686A111, TFIIIC, TFIIIC220, TFIIICalpha, TF3C-alpha; TFIIIC 220 kDa subunit; TFIIIC box B-binding subunit; general transcription factor 3C polypeptide 1; general transcription factor IIIC, polypeptide 1 (alpha subunit); transcription factor IIIC 220 kDa subunit; transcription factor IIIC subunit alpha
Gene Symbols GTF3C1
Host Species Rabbit
Immunogen Between 2059 and 2109
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2975
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.