Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540449 Supplier Invitrogen™ Supplier No. PA540449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YKADLIPYQD SPEGPDHSSA SSGVPNPPKA RTYTETVNHH YLLFQDTDLG Sequence homology: Cow: 85%; Dog: 92%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 77%; Rabbit: 85%; Rat: 85%.

Required for RNA polymerase III-mediated transcription. Component of TFIIIC that initiates transcription complex assembly on tRNA and is required for transcription of 5S rRNA and other stable nuclear and cytoplasmic RNAs. May play a direct role in stabilizing interactions of TFIIIC2 with TFIIIC1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3C2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GTF3C2
Gene Accession No. Q8WUA4
Gene Alias 1300004C11Rik; 2610510G03Rik; AI225816; AU041069; general transcription factor 3C polypeptide 2; general transcription factor IIIC subunit 2; general transcription factor IIIC, polypeptide 2, beta; general transcription factor IIIC, polypeptide 2, beta 110kDa; Gtf3c2; KIAA0011; mKIAA0011; TF3C-beta; TFIIIC 110 kDa subunit; TFIIIC110; TFIIIC-BETA; Transcription factor IIIC 110 kDa subunit; transcription factor IIIC subunit beta
Gene Symbols GTF3C2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GTF3C2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.