Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF3C2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557418
Description
Immunogen sequence: LRREPMLRMQ EGEGHSQLCL DRLQLEAIHK VRFSPNLDSY GWLVSGGQSG LVRIHFVRGL ASPLGHRMQL ESRAHFNAMF QPSSPTRRPG FSPTSH Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Required for RNA polymerase III-mediated transcription. Component of TFIIIC that initiates transcription complex assembly on tRNA and is required for transcription of 5S rRNA and other stable nuclear and cytoplasmic RNAs. May play a direct role in stabilizing interactions of TFIIIC2 with TFIIIC1. [UniProt].
Specifications
GTF3C2 | |
Polyclonal | |
Unconjugated | |
GTF3C2 | |
1300004C11Rik; 2610510G03Rik; AI225816; AU041069; general transcription factor 3C polypeptide 2; general transcription factor IIIC subunit 2; general transcription factor IIIC, polypeptide 2, beta; general transcription factor IIIC, polypeptide 2, beta 110kDa; Gtf3c2; KIAA0011; mKIAA0011; TF3C-beta; TFIIIC 110 kDa subunit; TFIIIC110; TFIIIC-BETA; Transcription factor IIIC 110 kDa subunit; transcription factor IIIC subunit beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2976 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUA4 | |
GTF3C2 | |
Recombinant protein corresponding to Human GTF3C2. Recombinant protein control fragment (Product #RP-96554). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction