Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF3C2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538747
Description
Required for RNA polymerase III-mediated transcription. Component of TFIIIC that initiates transcription complex assembly on tRNA and is required for transcription of 5S rRNA and other stable nuclear and cytoplasmic RNAs. May play a direct role in stabilizing interactions of TFIIIC2 with TFIIIC1.
Specifications
| GTF3C2 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C2 | |
| TFIIIC-BETA, TFIIIC110, TF3C-beta, TFIIIC 110 kDa subunit, general transcription factor 3C polypeptide 2, transcription factor IIIC 110 kDa subunit, transcription factor IIIC subunit beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WUA4 | |
| GTF3C2 | |
| A synthetic peptide derived from the internal region of human TF3C2 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction