Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF3C4 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060434AF405
Description
GTF3C4 Polyclonal antibody specifically detects GTF3C4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GTF3C4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.3.1.48, FLJ21002, general transcription factor 3C 4, general transcription factor 3C polypeptide 4, general transcription factor IIIC, polypeptide 4 (90kD), general transcription factor IIIC, polypeptide 4, 90kDa, KAT12, TF3C-delta, TFIII90, TFIIIC 90 kDa subunit, TFIIIC290, TFIIIC90MGC138450, TFIIICDELTA, Transcription factor IIIC 90 kDa subunit, Transcription factor IIIC subunit delta | |
The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human general transcription factor 3C polypeptide 4 (Transcription factor IIIC 90 kDa subunit) using the numbering given in entry NP_036336.1 (GeneID 9329). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9329 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction