Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF3C4 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA812629

Description
Essential for RNA polymerase III to make a number of small nuclear and cytoplasmic RNAs, including 5S RNA, tRNA, and adenovirus-associated (VA) RNA of both cellular and viral origin. Has histone acetyltransferase activity (HAT) with unique specificity for free and nucleosomal H3. May cooperate with GTF3C5 in facilitating the recruitment of TFIIIB and RNA polymerase through direct interactions with BRF1, POLR3C and POLR3F. May be localized close to the A box.Specifications
GTF3C4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UKN8 | |
GTF3C4 | |
Human recombit protein fragment corresponding to amino acids 583-822 of human GTF3C4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4A3 | |
Unconjugated | |
GTF3C4 | |
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA | |
Mouse | |
Affinity Chromatography | |
RUO | |
9329 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction