Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3C4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567046 Supplier Invitrogen™ Supplier No. PA567046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FWRFKLFLLRI LYQSMQKTPS EALWKPTHED SKILLVDSPG MGNADDEQQE EGTSSKQVVK QGLQERSKEG DVEEPTDDSL PT Highest antigen sequence identity to the following orthologs - mouse 82%, rat 81%.

Essential for RNA polymerase III to make a number of small nuclear and cytoplasmic RNAs, including 5S RNA, tRNA, and adenovirus-associated (VA) RNA of both cellular and viral origin. Has histone acetyltransferase activity (HAT) with unique specificity for free and nucleosomal H3. May cooperate with GTF3C5 in facilitating the recruitment of TFIIIB and RNA polymerase through direct interactions with BRF1, POLR3C and POLR3F. May be localized close to the A box. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3C4
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF3C4
Gene Accession No. Q9UKN8
Gene Alias 5330400C03; AI426938; AU014771; AU017413; general transcription factor 3C 4; general transcription factor 3C polypeptide 4; general transcription factor IIIC 4; general transcription factor IIIC subunit 4; general transcription factor IIIC, polypeptide 4; general transcription factor IIIC, polypeptide 4, 90kDa; Gtf3c4; KAT12; TF3C-delta; TFIII90; TFIIIC 90 kDa subunit; TFIIIC290; TFIIIC90; TFIIICDELTA; transcription factor IIIC 90 kDa subunit; Transcription factor IIIC subunit delta
Gene Symbols GTF3C4
Host Species Rabbit
Immunogen Recombinant Human GTF3C4. Recombinant protein control fragment (Product #RP-105475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9329
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.