Learn More
GTF3C4/TFIIIC90 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | GTF3C4/TFIIIC90 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1770
![]() |
Bethyl Laboratories, Inc
IHC00256T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1769
![]() |
Bethyl Laboratories, Inc
IHC00256 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee and Northern white-cheeked gibbon
Essential for RNA polymerase III to make a number of small nuclear and cytoplasmic RNAs, including 5S RNA, tRNA, and adenovirus-associated (VA) RNA of both cellular and viral origin. Has histone acetyltransferase activity (HAT) with unique specificity for free and nucleosomal H3. May cooperate with GTF3C5 in facilitating the recruitment of TFIIIB and RNA polymerase through direct interactions with BRF1, POLR3C and POLR3F. May be localized close to the A box.Specifications
GTF3C4/TFIIIC90 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
KAT12, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA, TF3C-delta, TFIIIC 90 kDa subunit, general transcription factor 3C 4, general transcription factor 3C polypeptide 4, transcription factor IIIC 90 kDa subunit, transcription factor IIIC subunit delta | |
GTF3C4 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9UKN8 | |
9329 | |
Between 625 and 675 | |
Primary | |
4° C | |
GTF3C4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.