Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3C5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541134 Supplier Invitrogen™ Supplier No. PA541134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KVLMLRPEKE AFFHQELPLY IPPPIFSRLD APVDYFYRPE TQHREGYNNP Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3C5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gtf3c5
Gene Accession No. Q9Y5Q8
Gene Alias 2700084A09Rik; CDABP0017; General transcription factor 3C polypeptide 5; general transcription factor III polypeptide 5; general transcription factor IIIC subunit 5; general transcription factor IIIC, polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; GTF3C5; im:7156509; LOW QUALITY PROTEIN: general transcription factor 3C polypeptide 5; TF3C-epsilon; TFIIIC 63 kDa subunit; TFiiiC2-63; TFIIIC63; TFIIICepsilon; transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD
Gene Symbols Gtf3c5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GTF3C5 (aa 148-197).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9328
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.