Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF3C5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564307 Supplier Invitrogen™ Supplier No. PA564307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HREGYNNPPI SGENLIGLSR ARRPHNAIFV NFEDEEVPKQ PLEAAAQTWR RVCTNPVDRK VEEELRKLFD IRPIWSRNA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF3C5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gtf3c5
Gene Accession No. Q9Y5Q8
Gene Alias 2700084A09Rik; CDABP0017; General transcription factor 3C polypeptide 5; general transcription factor III polypeptide 5; general transcription factor IIIC subunit 5; general transcription factor IIIC, polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; GTF3C5; im:7156509; LOW QUALITY PROTEIN: general transcription factor 3C polypeptide 5; TF3C-epsilon; TFIIIC 63 kDa subunit; TFiiiC2-63; TFIIIC63; TFIIICepsilon; transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD
Gene Symbols Gtf3c5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF3C5. Recombinant protein control fragment (Product #RP-103767).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.