Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF3C5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570368
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 77%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Yeast: 82%; Zebrafish: 86%.
Involved in RNA polymerase III-mediated transcription. Integral, tightly associated component of the DNA-binding TFIIIC2 subcomplex that directly binds tRNA and virus-associated RNA promoters. [UniProt].
Specifications
GTF3C5 | |
Polyclonal | |
Unconjugated | |
Gtf3c5 | |
2700084A09Rik; CDABP0017; General transcription factor 3C polypeptide 5; general transcription factor III polypeptide 5; general transcription factor IIIC subunit 5; general transcription factor IIIC, polypeptide 5; general transcription factor IIIC, polypeptide 5, 63kDa; GTF3C5; im:7156509; LOW QUALITY PROTEIN: general transcription factor 3C polypeptide 5; TF3C-epsilon; TFIIIC 63 kDa subunit; TFiiiC2-63; TFIIIC63; TFIIICepsilon; transcription factor IIIC 63 kDa subunit; Transcription factor IIIC subunit epsilon; transcription factor IIIC, 63 kD | |
Rabbit | |
Affinity chromatography | |
RUO | |
362095 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
Gtf3c5 | |
synthetic peptide directed towards the middle region of rat GTF3C5. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction