Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF3C5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | GTF3C5 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GTF3C5 Polyclonal specifically detects GTF3C5 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| GTF3C5 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ20857, general transcription factor 3C polypeptide 5, general transcription factor IIIC, polypeptide 5 (63kD), general transcription factor IIIC, polypeptide 5, 63kDa, TF3C-epsilon, TFiiiC2-63, TFIIIC63TFIIIC 63 kDa subunit, TFIIICepsilon, Transcription factor IIIC 63 kDa subunit, Transcription factor IIIC subunit epsilon, transcription factor IIIC, 63 kD | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human GTF3C5 (NP_036219). Peptide sequence SKRPALFSSSAKADGGKEQLTYESGEDEEDEEEEEEEEEDFKPSDGSENE | |
| Affinity purified |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 9328 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title