Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF3C5/TFIIIC63 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571772
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1772 10 μL
50-157-1771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1772 Supplier Bethyl Laboratories, Inc Supplier No. IHC00257T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Oranguta

GTF3C5 is polypeptide 5 of general transcription factor IIIon Channel. Human transcription factor IIIon Channel (hTFIIIon Channel) is a multisubunit complex that directly recognizes promoter elements and recruits TFIIIB and RNA polymerase III.

Specifications

Antigen GTF3C5/TFIIIC63
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GTF3C5
Gene Accession No. Q9Y5Q8
Gene Alias general transcription factor IIIC, polypeptide 5 (63kD), general transcription factor IIIC, polypeptide 5, 63kDa, TF3C-epsilon, TFIIIC 63 kDa subunit, TFiiiC2-63, TFIIIC63, TFIIICepsilon, transcription factor IIIC 63 kDa subunit, transcription factor IIIC subunit epsilon, transcription factor IIIC, 63 kD
Gene Symbols GTF3C5
Host Species Rabbit
Immunogen Between 469 and 519
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9328
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.