Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540859 Supplier Invitrogen™ Supplier No. PA540859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YGTNYIETLR VQIHANCRIR RVYFSDRLYS EDELPAEFKL YLPVQNKAKQ Sequence homology: Cow: 100%; Dog: 100%; Goat: 77%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

CFAP20 (Cilia And Flagella Associated Protein 20) is a Protein Coding gene. Diseases associated with CFAP20 includeFamilial Atrial FibrillationandMaturity-Onset Diabetes Of The Young, Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFAP20
Gene Accession No. Q9Y6A4
Gene Alias 2600014O15Rik; AL024127; Basal body up-regulated protein 22; BUG22; C16orf80; C18H16orf80; C2H16orf80; C3H16orf80; C5H16orf80; CFAP20; cilia and flagella associated protein 20; cilia- and flagella-associated protein 20; EVORF; flagellar associated protein 20 homolog; fSAP23; functional spliceosome-associated protein 23; gene trap locus 3; gene trap locus 3 protein; Gene trap locus 3 protein homolog; GTL3; likely ortholog of mouse gene trap locus 3; similar to transcription factor IIB; T10-2A2; transcription factor 2B; transcription factor IIB; UPF0468 protein C16orf80; UPF0468 protein C16orf80 homolog; zgc:73380
Gene Symbols CFAP20
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GTL3 (aa 144-193).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29105
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.