Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTP binding protein REM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987787
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7787 Supplier Bioss Supplier No. BS7502R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a GTPase and member of the RAS-like GTP-binding protein family. The encoded protein is expressed in endothelial cells, where it promotes reorganization of the actin cytoskeleton and morphological changes in the cells.

Specifications

Antigen GTP binding protein REM1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene REM1
Gene Accession No. O75628
Gene Alias BOS_13356; E030011C07Rik; GD:REM; GEM; GES; GTPase-regulating endothelial cell sprouting; GTP-binding protein REM 1; horizin; LOC100354515; rad and gem related GTP binding protein 1; Rad and Gem-like GTP-binding protein 1; Ras; RAS (RAD and GEM)-like GTP-binding 1; RAS-like GTP binding protein Rem; RAS-like GTP-binding protein REM; Rem; REM1; RRAD and GEM like GTPase 1; SI:zK76P14.3; zgc:56144
Gene Symbols REM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GTP binding protein REM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28954
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.