Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566710 Supplier Invitrogen™ Supplier No. PA566710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKLVLVSPTSE QYDSLLRQMW ERMDEGCGET IYVIGQGSDG TEYGLSEADM EASYATVKSM AEQIEADVIL LRERQ Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTPBP1
Gene Accession No. O00178
Gene Alias AL022987; AW045683; C85212; GP1; GP-1; G-protein 1; GTP binding protein 1; GTP-binding protein 1; Gtpbp; Gtpbp1; HSPC018; LOW QUALITY PROTEIN: GTP-binding protein 1
Gene Symbols GTPBP1
Host Species Rabbit
Immunogen Recombinant Human GTPBP1. Recombinant protein control fragment (Product #RP-105334).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.