Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115334 Supplier Invitrogen™ Supplier No. PA5115334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GTPBP1.

Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GTPBP1
Gene Accession No. D2XV59, O00178, O08582
Gene Alias AL022987; AW045683; C85212; GP1; GP-1; G-protein 1; GTP binding protein 1; GTP-binding protein 1; Gtpbp; Gtpbp1; HSPC018; LOW QUALITY PROTEIN: GTP-binding protein 1
Gene Symbols GTPBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GTPBP1(Accession O00178), corresponding to amino acid residues K422-S472.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14904, 300077, 9567
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.