Learn More
GTPBP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304662AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.Specifications
GTPBP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O00178 | |
GTPBP1 | |
Between 575 and 625 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GTPBP1 | |
GP1, GP-1, G-protein 1, HSPC018 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9567 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.