Learn More
GTPBP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304661A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.Specifications
| GTPBP1 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP1 | |
| GP1, GP-1, G-protein 1, HSPC018 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9567 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O00178 | |
| GTPBP1 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.