Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTPBP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554324
Description
Immunogen sequence: ALKGSKGKDC EIPVPVGISV TDENGKIIGE LNKENDRILV AQGGLGGKLL TNFLPLKGQK RIIHLDLKLI ADVGLVGFPN AGKSSLLSCV SHAKPAIADY AF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction.Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM].
Specifications
| GTPBP10 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP10 | |
| 4930545J22Rik; BC034507; claudin 12; Cldn12; DKFZP686A10121; FLJ38242; GTP binding protein 10; GTP binding protein 10 (putative); GTP-binding protein 10; GTP-binding protein 10 (putative); GTPBP10; MGC104191; ObgH2; Protein obg homolog 2; UG0751c10; Unknown (protein for MGC:128459); zgc:92334 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85865 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A4D1E9 | |
| GTPBP10 | |
| Recombinant protein corresponding to Human GTPBP10. Recombinant protein control fragment (Product #RP-92634). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction