Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542265 Supplier Invitrogen™ Supplier No. PA542265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IILLTKELEL YKEELQTKPA LLAVNKMDLP DAQDKFHELM SQLQNPKDFL Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction.Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP10
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GTPBP10
Gene Accession No. A4D1E9
Gene Alias 4930545J22Rik; BC034507; claudin 12; Cldn12; DKFZP686A10121; FLJ38242; GTP binding protein 10; GTP binding protein 10 (putative); GTP-binding protein 10; GTP-binding protein 10 (putative); GTPBP10; MGC104191; ObgH2; Protein obg homolog 2; UG0751c10; Unknown (protein for MGC:128459); zgc:92334
Gene Symbols GTPBP10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GTPBP10.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85865
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.