Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPBP10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985732
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5732 Supplier Bioss Supplier No. BS3893R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction.Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM].

Specifications

Antigen GTPBP10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GTPBP10
Gene Accession No. A4D1E9
Gene Alias 4930545J22Rik; BC034507; BOS_4364; claudin 12; Cldn12; DKFZP686A10121; FLJ38242; GTP binding protein 10; GTP binding protein 10 (putative); GTP-binding protein 10; GTP-binding protein 10 (putative); Gtpbp10; LOC100356629; MGC104191; OBGH2; Protein obg homolog 2; UG0751c10; zgc:92334
Gene Symbols GTPBP10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GTPBP10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85865
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.