Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTPBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556738
Description
Immunogen sequence: DGNIEYKLKL VNPSQYRFEH LVTQMKWRLQ EGRGEAVYQI GVEDNGLLVG LAEEEMRASL KTLHRMAEKV GADITVLRER EVDYDSDMPR KITEVLVRKV P Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
Small G proteins act as molecular switches for regulation of variety of cellular processes, such as nuclear transport, signal transduction, membrane trafficking and protein synthesis. GTPBP2 (GTP-binding protein 2) is a 602 amino acid G protein that is expressed in kidney, skeletal muscle, testis, brain and thymus, though it is not detected in liver. Expression of GTPBP2 is enhanced by ©-interferon stimulation in HeLa cells, THP-1 cells and thioglycollate-elicited mouse peritoneal macrophages. There are four isoforms of GTPBP2 that are expressed as a result of alternative splicing events. Since mutation of the gene encoding GTPBP1 does not lead to any phenotypic abnormalities, it is thought that there may be a genetic redundancy to make up for GTPBP1 lack-of-function. GTPBP2 shares 44% sequence similarity with GTPBP1 and also overlaps in expression pattern, suggesting that the GTPBP2 gene may compensate for GTPBP1 genetic abnormalities.
Specifications
GTPBP2 | |
Polyclonal | |
Unconjugated | |
Gtpbp2 | |
GTP binding protein 2; GTP-binding protein 2; GTP-binding-like protein 2; gtpbp2; I79_024595; nmf205 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54676 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BX10 | |
Gtpbp2 | |
Recombinant protein corresponding to Human GTPBP2. Recombinant protein control fragment (Product #RP-94951). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction