Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565097 Supplier Invitrogen™ Supplier No. PA565097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFQVDEIYTVP EVGTVVGGTL SSGICREGDQ LVVGPTDDGC FLELRVCSIQ RNRSACRVLR AGQAATLALG DFDRALLRKG MVMVSPEM Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Small G proteins act as molecular switches for regulation of variety of cellular processes, such as nuclear transport, signal transduction, membrane trafficking and protein synthesis. GTPBP2 (GTP-binding protein 2) is a 602 amino acid G protein that is expressed in kidney, skeletal muscle, testis, brain and thymus, though it is not detected in liver. Expression of GTPBP2 is enhanced by ©-interferon stimulation in HeLa cells, THP-1 cells and thioglycollate-elicited mouse peritoneal macrophages. There are four isoforms of GTPBP2 that are expressed as a result of alternative splicing events. Since mutation of the gene encoding GTPBP1 does not lead to any phenotypic abnormalities, it is thought that there may be a genetic redundancy to make up for GTPBP1 lack-of-function. GTPBP2 shares 44% sequence similarity with GTPBP1 and also overlaps in expression pattern, suggesting that the GTPBP2 gene may compensate for GTPBP1 genetic abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gtpbp2
Gene Accession No. Q9BX10
Gene Alias GTP binding protein 2; GTP-binding protein 2; GTP-binding-like protein 2; gtpbp2; I79_024595; nmf205
Gene Symbols Gtpbp2
Host Species Rabbit
Immunogen Recombinant Human GTPBP2. Recombinant protein control fragment (Product #RP-105855).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54676
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.