Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559895 Supplier Invitrogen™ Supplier No. PA559895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HALRILTAPR DLPLARHASL RLLSDPRSGE PLDRALVLWF PGPQSFTGED CVEFHVHGGP AVVSGVLQAL GSVPGLRPAE AGEFTRRAFA NGKLNL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

This locus encodes a GTP-binding protein. The encoded protein is localized to the mitochondria and may play a role in mitochondrial tRNA modification. Polymorphisms at this locus may be associated with severity of aminoglycoside-induced deafness, a disease associated with a mutation in the 12S rRNA. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTPBP3
Gene Accession No. Q969Y2
Gene Alias 2410009F13Rik; AI607903; COXPD23; GTP binding protein 3; GTP binding protein 3 (mitochondrial); GTPBG3; GTP-binding protein 3; Gtpbp3; Mitochondrial GTP-binding protein 1; MSS1; MTGP1; THDF1; tRNA modification GTPase GTPBP3, mitochondrial
Gene Symbols GTPBP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTPBP3. Recombinant protein control fragment (Product #RP-99071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84705
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.