Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTPBP4 Antibody (SR2224), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32204750UL
Description
GTPBP4 Monoclonal antibody specifically detects GTPBP4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
GTPBP4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
SR2224 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Chronic renal failure gene protein, CRFGGTP-binding protein NGB, FLJ10686, FLJ10690, G protein-binding protein CRFG, GTP binding protein 4, NGB, NOG1FLJ39774, nucleolar GTP-binding protein 1 | |
A synthesized peptide derived from human GTPBP4 (Uniprot #: Q9BZE4) | |
50 μg | |
Signal Transduction | |
23560 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction