Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTPBP4, Rabbit anti-Human, Clone: JE47-47, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP280462
Description
GTPBP4 Monoclonal specifically detects GTPBP4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GTPBP4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200Flow 1:50-1:100 | |
Chronic renal failure gene protein, CRFGGTP-binding protein NGB, FLJ10686, FLJ10690, G protein-binding protein CRFG, GTP binding protein 4, NGB, NOG1FLJ39774, nucleolar GTP-binding protein 1 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JE47-47 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
GTPBP4 | |
Recombinant protein within Human GTPBP4 aa 1-131/ 634. (SwissProt: Q9BZE4 Human) | |
100 μL | |
Signal Transduction | |
23560 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction