Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP4 Recombinant Rabbit Monoclonal Antibody (JE47-47)
GREENER_CHOICE

Catalog No. PIMA534853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534853 Supplier Invitrogen™ Supplier No. MA534853
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: 293T cell, human liver cancer tissue, human thyroid gland tissue, human skin tissue, human breast tissue, thp-1.

GTP-binding proteins are GTPases and function as molecular switches that can flip between two states: active, when GTP is bound, and inactive, when GDP is bound. 'Active' in this context usually means that the molecule acts as a signal to trigger other events in the cell. When an extracellular ligand binds to a G-protein-linked receptor, the receptor changes its conformation and switches on the trimeric G proteins that associate with it by causing them to eject their GDP and replace it with GTP. The switch is turned off when the G protein hydrolyzes its own bound GTP, converting it back to GDP. But before that occurs, the active protein has an opportunity to diffuse away from the receptor and deliver its message for a prolonged period to its downstream target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE47-47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Gtpbp4
Gene Accession No. Q9BZE4
Gene Alias 2610028C09Rik; chronic renal failure gene protein; chronic renal failure protein; Crfg; fi28d07; FLJ10686; FLJ10690; FLJ39774; G protein-binding protein CRFG; GTP binding protein 3; GTP binding protein 4; GTP binding protein NGB; GTP-binding protein NGB; Gtpbp3; Gtpbp4; NGB; Nog1; nucleolar GTP-binding protein 1; Nucleolar GTP-binding protein 1 (Chronic renal failure gene protein) (GTP-binding protein NGB); Unknown (protein for IMAGE:7956347); wu:fi28d07; zgc:55757
Gene Symbols Gtpbp4
Host Species Rabbit
Immunogen Recombinant protein within Human GTPBP4 aa 1-131.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23560
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.