Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112829
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112829 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112829 Supplier Invitrogen™ Supplier No. PA5112829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTPBP6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GTPBP6
Gene Accession No. O43824
Gene Alias AV119224; GTP binding protein 6 (putative); Gtpbp6; Pgbpll; pgpl; pseudoautosomal GTP binding protein-like, like; Pseudoautosomal GTP-binding protein-like; Putative GTP-binding protein 6; RGD1305954
Gene Symbols GTPBP6
Host Species Rabbit
Immunogen Recombinant Human Putative GTP-binding protein 6 protein (146-295AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8225
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.