Learn More
Invitrogen™ GTPBP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116828
Description
Positive Samples: Mouse thymus, Rat thymus, Rat spleen Immunogen sequence: MVVSTKTPDR KLIFGKGNFE HLTEKIRGSP DITCVFLNVE RMAAPTKKEL EAAWGVEVFD RFTVVLHIFR CNARTKEARL QVALAEMPLH RSNLKRDVAH LYRGVGSRYI MGSGESFMQL QQRLLREKEA KIRKALDRLR KKRHLLRRQR TRREFPVISV VGYTNCGKTT LIKALTGDAA IQPRDQLFAT LDVTAHAGTL PSRMTVLYVD T.
Specifications
| GTPBP6 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP6 | |
| AV119224; GTP binding protein 6 (putative); Gtpbp6; Pgbpll; pgpl; pseudoautosomal GTP binding protein-like, like; Pseudoautosomal GTP-binding protein-like; Putative GTP-binding protein 6; RGD1305954 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107999, 363931, 8225 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O43824, Q3U6U5 | |
| GTPBP6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-350 of human GTPBP6 (NP_0363593). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.