Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584389 Supplier Invitrogen™ Supplier No. PA584389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIKALFSLAP EVEVRVSKKP GHTKKMNFFK VGKHFTVVDM PGYGFRAPED FVDMVETYLK ERRNL Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 92%.

GTPBP8 is a protein coding gene. Gene ontology (GO) annotation include mitochondrion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP8
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTPBP8
Gene Accession No. Q8N3Z3
Gene Alias GTP binding protein 8 (putative); GTP-binding protein 8; GTP-binding protein 8 (putative); GTPBP8; HSPC135
Gene Symbols GTPBP8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTPBP8. Recombinant protein control fragment (Product #RP-106876).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.