Learn More
Invitrogen™ GTPBP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531227
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse testis, rat testis. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (89%), Xenopus laevis (85%), Chicken (90%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hydrolyzes ATP, and can also hydrolyze GTP with lower efficiency. Has lower affinity for GTP.
Specifications
| GTPBP9 | |
| Polyclonal | |
| Unconjugated | |
| OLA1 | |
| 2510025G09Rik; 2810405J23Rik; 2810409H07Rik; DNA damage-regulated overexpressed in cancer 45; DOC45; fe36h01; GBP45; GTBP9; GTP-binding protein 9; GTP-binding protein 9 (putative); GTP-binding protein PTD004; Gtpbp9; HAMAP-Rule:MF_03167}; Obg like ATPase 1; Obg-like ATPase 1; obg-like ATPase 1 {ECO:0000255; obg-like ATPase 1; LOW QUALITY PROTEIN: obg-like ATPase 1; obg-like ATPase 1-like protein; OLA1; PRO2455; PTD004; RGD1307982; similar to Putative GTP-binding protein PTD004; Unknown (protein for MGC:55768); wu:fc66b09; wu:fe36h01; wu:fk82a02; zgc:55768; zgc:55768 protein; zgc:85691 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 296488, 29789, 67059 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.46 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| A0JPJ7, Q9CZ30, Q9NTK5 | |
| OLA1 | |
| Recombinant protein encompassing a sequence within the center region of human GTPBP9. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.