Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTPBP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531228 Supplier Invitrogen™ Supplier No. PA531228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, mouse testis, rat testis. Predicted reactivity: Mouse (99%), Rat (97%), Zebrafish (83%), Xenopus laevis (85%), Chicken (94%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hydrolyzes ATP, and can also hydrolyze GTP with lower efficiency. Has lower affinity for GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTPBP9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene OLA1
Gene Accession No. A0JPJ7, Q9CZ30, Q9NTK5
Gene Alias 2510025G09Rik; 2810405J23Rik; 2810409H07Rik; DNA damage-regulated overexpressed in cancer 45; DOC45; fe36h01; GBP45; GTBP9; GTP-binding protein 9; GTP-binding protein 9 (putative); GTP-binding protein PTD004; Gtpbp9; HAMAP-Rule:MF_03167}; Obg like ATPase 1; Obg-like ATPase 1; obg-like ATPase 1 {ECO:0000255; obg-like ATPase 1; LOW QUALITY PROTEIN: obg-like ATPase 1; obg-like ATPase 1-like protein; OLA1; PRO2455; PTD004; RGD1307982; similar to Putative GTP-binding protein PTD004; Unknown (protein for MGC:55768); wu:fc66b09; wu:fe36h01; wu:fk82a02; zgc:55768; zgc:55768 protein; zgc:85691
Gene Symbols OLA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GTPBP9. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296488, 29789, 67059
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.