Learn More
GTPBP9 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 163711AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0899
Only null left
Add to Cart
Description
Description
SLC7A1 is a high-affinity, low capacity permease involved in the transport of the cationic amino acids (arginine, lysine and ornithine) in non-hepatic tissues. It may also function as an ecotropic retroviral leukemia receptor.
Specifications
Specifications
GTPBP9
Polyclonal
Unconjugated
OLA1
GTP binding protein 9, GTPBP9, Obg like ATPase 1, OLA1, PRO2455, PTD004
Rabbit
Antigen Affinity Chromatography
RUO
29789
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NTK5
OLA1
GTPBP9 Fusion Protein Ag9283
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.