Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTSE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103414
Description
Antibody detects endogenous levels of total GTSE1.
The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.
Specifications
GTSE1 | |
Polyclonal | |
Unconjugated | |
GTSE1 | |
B99; G two S phase expressed protein 1; G2 and S phase-expressed protein 1; G-2 and S-phase expressed 1; G2 and S-phase expressed 1; GTSE1; GTSE-1; Protein B99; Protein B99 homolog; RGD1563164 | |
Rabbit | |
Affinity chromatography | |
RUO | |
29870, 51512 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R080, Q9NYZ3 | |
GTSE1 | |
A synthesized peptide derived from human GTSE1(Accession Q9NYZ3), corresponding to amino acid residues R380-I430. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction