Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109311 Supplier Invitrogen™ Supplier No. PA5109311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Zebrafish.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GTSF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene Alias 1700006H03Rik; Cue110; Fam112b; family with sequence similarity 112, member B; gametocyte specific factor 1; gametocyte-specific factor 1; Gtsf1; Protein FAM112B; RGD1310791
Host Species Rabbit
Immunogen Recombinant protein within zebrafish GTSF1 aa 1-182.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.