Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116852 Supplier Invitrogen™ Supplier No. PA5116852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Rat brain Immunogen sequence: MEETYTDSLD PEKLLQCPYD KNHQIRACRF PYHLIKCRKN HPDVASKLAT CPFNARHQVP RAEISHHISS CDDRSCIEQD VVNQTRSLRQ ETLAESTWQC PPCDEDWDKD LWEQTSTPFV WGTTHYSDNN SPASNIVTEH KNNLASGMRV PKSLPYVLPW KNNGNAQ.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GTSF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GTSF1
Gene Accession No. Q8WW33, Q9DAN6
Gene Alias 1700006H03Rik; Cue110; Fam112b; family with sequence similarity 112, member B; gametocyte specific factor 1; gametocyte-specific factor 1; Gtsf1; Protein FAM112B; RGD1310791
Gene Symbols GTSF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human GTSF1 (NP_6531952).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121355, 315347, 74174
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.