Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiac Tincture 1:60, Ricca Chemical
SDP

Catalog No. p-3251733
Click to view available options
Quantity:
120 mL

Specifications

Chemical Name or Material Guaiac Tincture
CAS Min % 81.0, 5.93, 4.3, 4.0, 1.98
CAS Max % 85.0, 6.17, 4.48, 6.0, 2.06
CAS 7732-18-5, 67-63-0, 67-56-1, 9000-29-7
Concentration 1:60
Packaging Natural Poly Bottle
Color Colorless
Quantity 120 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.