Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiazulene, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
489-84-9
C15H18
198.31
FWKQNCXZGNBPFD-UHFFFAOYSA-N
guaiazulene,1,4-dimethyl-7-isopropylazulene,azulon,vetivazulen,azunol,7-isopropyl-1,4-dimethylazulene,eucazulen,guajazulene,kessazulen,purazulen
3515
CHEBI:5550
1,4-dimethyl-7-propan-2-ylazulene
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C
489-84-9
C15H18
198.31
FWKQNCXZGNBPFD-UHFFFAOYSA-N
guaiazulene,1,4-dimethyl-7-isopropylazulene,azulon,vetivazulen,azunol,7-isopropyl-1,4-dimethylazulene,eucazulen,guajazulene,kessazulen,purazulen
3515
CHEBI:5550
1,4-dimethyl-7-propan-2-ylazulene
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C
Specific Gravity 0.97
Quantity 10g
Melting Point 29.0°C to 31.0°C
Infrared Spectrum Authentic
Flash Point >110°C
Density 0.9700g/mL
Boiling Point 153.0°C (7.0 Torr)
Packaging Glass bottle
Color Blue
Assay Percent Range 99%
Beilstein 05,II,473
Merck Index 15,459
Solubility Information Solubility in water: 25%. Other solubilities: soluble in ethanol, ethyl ether and ethyl acetate
Formula Weight 198.31
Physical Form Low Melting Crystalline Solid
Percent Purity 99%
Chemical Name or Material Guaiazulene
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.